Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXW9 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297608AF405
Description
FBXW9 Polyclonal specifically detects FBXW9 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
FBXW9 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
F-box and WD repeat domain containing 9, F-box and WD-40 domain protein 9, F-box and WD-40 domain-containing protein 9, F-box and WD-40 repeat containing protein 9, specificity factor for SCF ubiquitin ligase | |
Produced in rabbits immunized with E. coli-derived Human FBXW9 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
84261 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction