Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXW9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | FBXW9 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FBXW9 Polyclonal specifically detects FBXW9 in Human samples. It is validated for Western Blot.Specifications
| FBXW9 | |
| Polyclonal | |
| Rabbit | |
| NP_115677 | |
| 84261 | |
| Synthetic peptide directed towards the C terminal of human FBXW9The immunogen for this antibody is FBXW9. Peptide sequence PDILVTGTYDKKVTIYDPRAGPALLKHQQLHSRPVLTLLADDRHIISGSE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| F-box and WD repeat domain containing 9, F-box and WD-40 domain protein 9, F-box and WD-40 domain-containing protein 9, F-box and WD-40 repeat containing protein 9, specificity factor for SCF ubiquitin ligase | |
| FBXW9 | |
| IgG | |
| 51 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title