Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXW9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570482
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 79%; Rat: 100%.
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex.
Specifications
FBXW9 | |
Polyclonal | |
Unconjugated | |
FBXW9 | |
1110017H11Rik; F-box and WD repeat domain containing 9; F-box and WD-40 domain protein 9; F-box and WD-40 domain-containing protein 9; F-box and WD-40 repeat containing protein 9; F-box/WD repeat-containing protein 9; Fbw9; FBXW9; specificity factor for SCF ubiquitin ligase; Unknown (protein for MGC:134310) | |
Rabbit | |
Affinity chromatography | |
RUO | |
84261 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5XUX1 | |
FBXW9 | |
synthetic peptide directed towards the C terminal region of human FBXW9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction