Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXW9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559862
Description
Immunogen sequence: VEEKNFDWPA ACIALEQHLS RWAEDGRWVE YFCLAEGHVA SVDSVLLLQG GSLCLSGSRD RNVNLWDLRQ LGTESNQVLI Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex.
Specifications
FBXW9 | |
Polyclonal | |
Unconjugated | |
FBXW9 | |
1110017H11Rik; F-box and WD repeat domain containing 9; F-box and WD-40 domain protein 9; F-box and WD-40 domain-containing protein 9; F-box and WD-40 repeat containing protein 9; F-box/WD repeat-containing protein 9; Fbw9; FBXW9; specificity factor for SCF ubiquitin ligase; Unknown (protein for MGC:134310) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84261 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5XUX1 | |
FBXW9 | |
Recombinant protein corresponding to Human FBXW9. Recombinant protein control fragment (Product #RP-99098). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction