Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc epsilon RI Antibody (9E1), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063269AF350
Description
Fc epsilon RI Monoclonal specifically detects Fc epsilon RI in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Fc epsilon RI | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
FCER1A | |
Made to Human Fc EPSILON R1 ALPHA | |
Primary | |
NB100-63269 recognizes the human high affinity Fc receptor for IgE (Fc epsilon R1), which plays a central role in the IgE-mediated allergic response. Fc epsilon R1 consists of four subunits, a high affinity IgE binding alpha subunit, a beta chain and two di-sulphide linked gamma subunits. Clone 9E1 specifically recognizes the extracellular D1 domain of the Fc epsilon R1 alpha chain. Fc epsilon R1 is primarily expressed on mast cells and basophils but expression of Fc epsilon R1 has also been reported on monocytes, Langerhans cells and dendritic cells from patients with atopic diseases. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
90 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide | |
Mouse | |
0.1 mL | |
2205 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction