Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc epsilon RI Antibody (IB10), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312050UV
Description
Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
Fc epsilon RI | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG κ |
Flow Cytometry, Immunofluorescence | |
IB10 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide | |
Recombinant human protein, N-terminus. | |
0.1 mL | |
Apoptosis, Asthma, Cancer, Immunology, Signal Transduction | |
2205 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction