Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc epsilon RI Mouse anti-Human, Clone: IE7, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | Fc epsilon RI |
---|---|
Clone | IE7 |
Dilution | Flow Cytometry |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Human samples. It is validated for Flow CytometrySpecifications
Fc epsilon RI | |
Flow Cytometry | |
Monoclonal | |
Mouse | |
Apoptosis, Asthma, Cancer, Immunology, Signal Transduction | |
PBS | |
2205 | |
IgG1 κ | |
Protein A purified |
IE7 | |
Flow Cytometry | |
Unconjugated | |
RUO | |
Human | |
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide | |
Recombinant human protein, N-terminus. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title