Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc epsilon RI Rabbit anti-Human, Clone: IE7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP3120520.2MG
Description
Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
Fc epsilon RI | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide | |
Recombinant human protein, N-terminus. | |
0.2 mg | |
Apoptosis, Asthma, Cancer, Immunology, Signal Transduction | |
2205 | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Immunofluorescence | |
IE7 | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction