Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc Epsl R1 Alp Mouse, anti-Human, R718, Clone: AER-37, BD OptiBuild™

Catalog No. BDB752192 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752192 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752192 Supplier BD Biosciences Supplier No. 752192
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The AER-37 (CRA-1) monoclonal antibody specifically recognizes FcεRIα which is also known as the high affinity immunoglobulin epsilon receptor subunit alpha (Fc-epsilon RI-alpha). FcεRIα is a type I transmembrane glycoprotein that is encoded by FCER1A (Fc fragment of IgE receptor Ia) which belongs to the Ig gene superfamily. FcεRIα serves as the binding subunit for the Fc region of IgE. It forms part of a heterotetrameric, high-affinity IgE Fc receptor (FceR1/FcεR1) that includes signal transducing subunits, one β-chain (FcεRIβ encoded by MS4A2) and two disulfide-linked γ-subunits (FcεRIγ encoded by FCER1G). FcεRIα is normally expressed on basophils and mast cells and can also be expressed on some monocytes, Langerhans cells, dendritic cells, and eosinophils from allergic donors. FcεRIα plays a major role in allergic responses and in the presentation of allergens to the immune system. The AER-37 antibody reportedly does not compete with IgE for FceR1 binding.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc Epsl R1 Alp
Applications Flow Cytometry
Classification Monoclonal
Clone AER-37
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.