Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RI/CD64 Antibody (005), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290842AF647
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Rat samples. It is validated for Flow Cytometry.Specifications
| Fc gamma RI/CD64 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Rat Fc gamma RI/CD64 (NP_001094306.1; Met1-Pro285). | |
| 0.1 mL | |
| Primary | |
| Rat | |
| Purified |
| Flow Cytometry | |
| 5 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2209 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction