Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RI/CD64 Antibody (276426) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB12571J
Description
Fc gamma RI/CD64 Monoclonal antibody specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
Fc gamma RI/CD64 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
276426 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
Mouse myeloma cell line NS0-derived recombinant human Fc gamma RI, Gln16-Pro288, Accession # P12314.2 | |
0.1 mL | |
CD Markers, Immunology | |
2209 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction