Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RI/CD64 Antibody (407), Alexa Fluor™ 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289363AF350
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
Fc gamma RI/CD64 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
2209 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, ELISA | |
407 | |
Flow Cytometry, ELISA | |
CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Fc gamma RI/CD64 (NP_000557.1; Met1-Pro288). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction