Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RI/CD64 Antibody (H22), Alexa Fluor™ 647, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP281097AF647
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Competitive ELISA.Specifications
Fc gamma RI/CD64 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
FCGR1A | |
This recombinant antibody was prepared by cloning the single chain fragment (scFv) derived from the murine anti-human Fc gamma RI/CD64 mAb m22 into the bacterial expression vector pBM1.1, and fusing it to a deletion mutant of Pseudomonas exotoxin A. | |
0.1 mL | |
Primary | |
This immunotoxin is specific for human Fc gamma RI/CD64, and specifically an epitope outside the Fc binding domain. The binding activity of the VH/VL antibody format was unaffected by fusion of the m22(scFv) coding regions to the truncated ETA coding sequences | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
H22 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Competitive ELISA | |
CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
Human | |
Protein A purified | |
RUO | |
2209 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction