Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RI/CD64 Antibody (OTI1A8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270702H
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Fc gamma RI/CD64 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
FCGR1A | |
Full length recombinant protein of human Fc gamma RI/CD64 (NP_000557) produced in HEK293T cells. | |
0.1 mL | |
2209.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1A8 | |
Western Blot, Immunohistochemistry | |
CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction