Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RI/CD64 Rabbit anti-Human, Clone: 407, Novus Biologicals™

Rabbit Monoclonal Antibody
$524.00
Specifications
Antigen | Fc gamma RI/CD64 |
---|---|
Clone | 407 |
Dilution | Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
Fc gamma RI/CD64 | |
Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
2209 | |
IgG | |
Protein A purified |
407 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
Recombinant Human Fc gamma RI/CD64 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title