Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RII/CD32 Antibody (MM0270- 3L1), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212161AF405
Description
Fc gamma RII/CD32 Monoclonal specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
Fc gamma RII/CD32 | |
Monoclonal | |
Alexa Fluor 405 | |
P31994 | |
FCGR2B | |
Human recombinant Fc gamma RII/CD32 (extracellular domain) | |
0.1 mL | |
Immunology | |
2213 | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry | |
MM0270- 3L1 | |
Western Blot, Flow Cytometry, Immunocytochemistry | |
CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects human Fc gamma RIIa (CD32a) and Fc gamma RIIb (CD32b) | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction