Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RII/CD32 Antibody (MM0270- 3L1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
| Antigen | Fc gamma RII/CD32 |
|---|---|
| Clone | MM0270- 3L1 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Fc gamma RII/CD32 Monoclonal specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| Fc gamma RII/CD32 | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| P31994 | |
| 2213 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0270- 3L1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| FCGR2B | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title