Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RII/CD32 Antibody (SR1608), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32256550UL
Description
Fc gamma RII/CD32 Monoclonal antibody specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Fc gamma RII/CD32 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1608 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
A synthesized peptide derived from human Fc gamma RII/CD32 (Uniprot #: P12318, P31994, P31995) | |
50 μg | |
Immunology | |
2213 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction