Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Fc gamma RII/RIII (CD32/CD16) Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB1460V100UG
This item is not returnable.
View return policy
Description
Fc gamma RII/RIII (CD32/CD16) Monoclonal specifically detects Fc gamma RII/RIII (CD32/CD16) in Mouse samples. It is validated for Flow Cytometry.Specifications
Fc gamma RII/RIII (CD32/CD16) | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P08101 | |
Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIIB Thr30-Arg207 Accession # P08101 | |
RUO | |
2214 | |
Mouse | |
IgG2b |
Flow Cytometry | |
190909 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Rat | |
100 μg | |
Primary | |
Detects mouse Fc gamma RII/RIII (CD32/CD16) in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse CD16 is observed and no cross-reactivity with recombinant human CD32 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction