Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIIA/CD32a Antibody (7.3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | Fc gamma RIIA/CD32a |
---|---|
Clone | 7.3 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247830A
![]() |
Novus Biologicals
NBP2478300.1MG |
0.1 mg |
Each for $646.00
|
|
|||||
NBP247830B
![]() |
Novus Biologicals
NBP2478300.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Fc gamma RIIA/CD32a Monoclonal specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional, Immunofluorescence, CyTOF-ready.Specifications
Fc gamma RIIA/CD32a | |
1.0 mg/mL | |
Flow Cytometry, Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a | |
FCGR2A | |
IgG1 κ | |
Protein A or G purified | |
This MAb reacts with a CD32 (FcgRII) epitope (cluster-4). It displays a stronger reaction with Daudi than with U937 cells. The epitope is located in domain 2 of FcgRIIa. Its Fab'2 fragments block immune complex binding. CD32 (Fc?RII) is a type 1 transmembrane glycoprotein that mediates several functions including phagocytosis, cytotoxicity, and immunomodulation as well as platelet aggregation. Three genes (A, B, and C) encode CD32 and at least 6 isoforms are generated via alternative mRNA splicing, i.e., IIa1, IIa2, IIb1, IIb2, IIb3 and IIc. Monocytes/macrophages, placental trophoblasts and endothelial cells express all isoforms. In addition, the IIb isoform is expressed by B cells, and the IIa isoform by platelets, granulocytes and, weakly, by B cells. NK cells and neutrophils express Isoform IIc. CD32 binds weakly to the Fc region of monomeric IgG but more strongly to IgG aggregates and immune complexes. |
7.3 | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
2212 | |
K562 and FcgRII+L cells | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
40 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title