Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIIA/CD32a Antibody (8.7), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247831AF350
Description
Fc gamma RIIA/CD32a Monoclonal specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence.Specifications
Fc gamma RIIA/CD32a | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
FCGR2A | |
K562 and FcgRII+L cells | |
Immunology | |
2212 | |
Human | |
IgG1 κ |
Flow Cytometry, Immunofluorescence | |
8.7 | |
Flow Cytometry, Immunofluorescence | |
CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a | |
Mouse | |
0.1 mL | |
Primary | |
This monoclonal antibody reacts with a CD32 (FcgRII) epitope distinct from that defined by monoclonal antibody 8.26 and the epitope overlaps with that of monoclonal antibody 7.30 (cluster 4). It displays a stronger reaction with Daudi than with U937 cells. The epitope is located in domain 2 of FcgRIIa. Its Fab'2 fragments block immune complex binding. CD32 (FcRII) is a type 1 transmembrane glycoprotein that mediates several functions including phagocytosis, cytotoxicity, and immunomodulation as well as platelet aggregation. Three genes (A, B, and C) encode CD32 and at least 6 isoforms are generated via alternative mRNA splicing, i.e., IIa1, IIa2, IIb1, IIb2, IIb3 and IIc. Monocytes/macrophages, placental trophoblasts and endothelial cells express all isoforms. In addition, the IIb isoform is expressed by B cells, and the IIa isoform by platelets, granulocytes and, weakly, by B cells. NK cells and neutrophils express Isoform IIc. CD32 binds weakly to the Fc region of monomeric IgG but more strongly to IgG aggregates and immune complexes. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction