Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIIB/CD32b Antibody (110), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289364FR
Description
Fc gamma RIIB/CD32b Monoclonal specifically detects Fc gamma RIIB/CD32b in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Fc gamma RIIB/CD32b | |
Monoclonal | |
DyLight 680 | |
CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Fc gamma RIIB/CD32b extracellular domain (Ala 46-Pro 217; NP_001002274.1). | |
0.1 mL | |
Immunology, Phospho Specific | |
2213 | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
110 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human CD64 Human CD16b No cross-reactivity in ELISA with: Human CD32a Human CD16a Mouse FCGR2B | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction