Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIIB/CD32b Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299136PCP
Description
Fc gamma RIIB/CD32b Polyclonal antibody specifically detects Fc gamma RIIB/CD32b in Human samples. It is validated for ELISASpecifications
Fc gamma RIIB/CD32b | |
Polyclonal | |
ELISA | |
CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
Produced in rabbits immunized with purified, recombinant Human Fc gamma RIIB/CD32b (Accession#: Ala 46-Pro 217; NP_001002274.1) | |
0.1 mL | |
Immunology, Phospho Specific | |
2213 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction