Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIII (CD16) Antibody (015), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290536AF750
Description
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Western Blot, ELISA.Specifications
Fc gamma RIII (CD16) | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
FCGR3A | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fc gamma RIII (CD16) (P08508-1; Met 1-Thr 215). | |
0.1 mL | |
Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
2214 | |
Mouse | |
Purified |
Western Blot, ELISA | |
15 | |
Western Blot, ELISA | |
CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human CD64 No cross-reactivity in ELISA with: Human CD32a Human CD32b Human CD16a Human CD16b Cynomolgus FCGR3 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction