Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIII (CD16) Antibody (221), Alexa Fluor™ 700

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289461AF700
Description
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Flow Cytometry.Specifications
Fc gamma RIII (CD16) | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
FCGR3A | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Fc gamma RIII (CD16) (P08637-1; Met1-Gln208). | |
0.1 mL | |
Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
2214 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
221 | |
Flow Cytometry | |
CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction