Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIII (CD16) Antibody (ICO-116), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | Fc gamma RIIIA/CD16a |
---|---|
Clone | ICO-116 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24483800
![]() |
Novus Biologicals
NBP2448380.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP24483801
![]() |
Novus Biologicals
NBP2448380.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP24483802
![]() |
Novus Biologicals
NBP2448380.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence.Specifications
Fc gamma RIIIA/CD16a | |
0.2 mg/mL | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Astrocyte Markers, Cellular Markers, Cytokine Research, Cytoskeleton Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA | |
FCGR3A | |
IgG1 κ | |
Protein A or G purified | |
It recognizes CD16 (Fcgamma RIII), the low-affinity receptor for IgG with an apparent molecular weight of 50-80kDa. Two similar genes represent CD16, CD16A (Fcgamma RIIIA), which exists as a hetero-oligomeric polypeptide-anchored form in macrophages and NK cells and CD16B (Fcgamma RIIIB), which exist as a monomeric GPI-anchored form in neutrophils. Furthermore, there are two known polymorphisms of CD16B, NA-1 and NA-2. Individuals homozygous for NA-2 show a lower phagocytic capacity compared with NA-1. CD16 binds IgG in the form of immune complexes and shows preferential binding of IgG1 and IgG3 isotypes and minimal binding of IgG2 and IgG4. Upon IgG binding, both CD16 isoforms initiate signal transduction cascades that lead to a variety of responses including antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, degranulation and proliferation. |
ICO-116 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P08637 | |
2214 | |
Human CD16 protein | |
Primary | |
Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title