Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Fc gamma RIII (CD16) Antibody Pair [HRP]
SDP

Catalog No. NB010357 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010357 15 Plates
NB010358 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010357 Supplier Novus Biologicals™ Supplier No. NBP27965915PLATES
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rhesus Macaque Fc gamma RIII (CD16).

Specifications

Assay Range 12.5 pg/mL
Assay Sensitivity 12.5-800 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rhesus Macaque Fc gamma RIII (CD16).
Target Species Rhesus Macaque
For Use With (Application) Sandwich ELISA
Gene Alias CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
Gene ID (Entrez) 2214
Gene Symbol FCGR3A
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.25 mg/mL mouse anti-Fc gamma RIII (CD16) monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.07 mg/mL of mouse anti-Fc gamma RIII (CD16) monoclonal antibody.
  • Standard: Each vial contains 31 ng of recombinant Fc gamma RIII (CD16).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target Fc gamma RIII (CD16)
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.8 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.