Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIII (CD16) Rabbit anti-Human, Clone: 221, Novus Biologicals™

Rabbit Monoclonal Antibody
Specifications
Antigen | Fc gamma RIII (CD16) |
---|---|
Clone | 221 |
Dilution | Flow Cytometry 1:25-1:100 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB020406
![]() |
Novus Biologicals
NBP28946150UL |
50 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Flow Cytometry.Specifications
Fc gamma RIII (CD16) | |
Flow Cytometry 1:25-1:100 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
2214 | |
IgG | |
Protein A purified |
221 | |
Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human | |
CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
Recombinant Human Fc gamma RIII (CD16) Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title