Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIIIA/CD16a Antibody (2829B), Allophycocyanin/Cy7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB107512APCCY7
Description
Fc gamma RIIIA/CD16a Monoclonal antibody specifically detects Fc gamma RIIIA/CD16a in Human samples. It is validated for Flow CytometrySpecifications
Fc gamma RIIIA/CD16a | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2829B | |
Flow Cytometry | |
CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA | |
Mouse myeloma cell line NS0-derived human Fc gamma RIIIA/CD16a, Gly17-Gln208, Accession # P08637 | |
0.1 mL | |
Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
2214 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction