Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIIIA/CD16a Antibody (3G8), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281102JF646
Description
Fc gamma RIIIA/CD16a Monoclonal specifically detects Fc gamma RIIIA/CD16a in Human, Baboon, Chimpanzee, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Fc gamma RIIIA/CD16a | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
FCGR3A | |
3G8 was prepared by immunizing mice with human PMN (polymorphonuclear leukocytes) cells. | |
0.1 mL | |
Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
2214 | |
Human, Baboon, Chimpanzee, Cynomolgus Monkey, Rhesus Macaque | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
3G8 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
3G8 binds to CD16 at an epitope which is not the IgG-binding site. This antibody cannot distinguish between CD16a transmembrane isoform found on the surface of NK cells and macrophages (which can trigger the cytolytic response), and the GPI-linked CD16b isoform present on neutrophils or soluble CD16 found in the plasma. CD16 is the low affinity IgG receptor III and binds to aggregated IgG or IgG-antigen complex which functions in NK cell activation, phagocytosis and ADCC (antibody-dependent cell-mediated cytotoxicity). | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction