Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIIIA/CD16a Antibody (DJ130c), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064346AF350
Description
Fc gamma RIIIA/CD16a Monoclonal specifically detects Fc gamma RIIIA/CD16a in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Fc gamma RIIIA/CD16a | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FCGR3A | |
| CD34 antigen | |
| Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
| 2214 | |
| Human | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DJ130c | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| NB100-64346 recognizes human CD16, a 50-65kD cell surface molecule, which is the low affinity receptor for IgG (FcR III). CD16 exists as a transmembranous form (Fc gammaRIIIA, or CD16A) and a glycosyl phosphatidylinositol (GPI) anchored form (Fc gammaRIIIB, or CD16B). CD16A is expressed by NK cells, some T cells, and macrophages, whereas CD16B is primarily expressed by granulocytes. In addition, CD16B exists as two allelic variants NA1 and NA2. DJ130c recognizes all polymorphonuclear cells irrespective on their NA phenotype. Studies show that the DJ130c antibody recognizes an epitope in the first membrane-distal domain of CD16 and recognizes both CD16A and CD16B. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction