Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RIIIA/CD16a Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23375325UL
Description
Fc gamma RIIIA/CD16a Polyclonal specifically detects Fc gamma RIIIA/CD16a in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Fc gamma RIIIA/CD16a | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
P08637 | |
FCGR3A | |
This antibody was developed against a recombinant protein corresponding to amino acids: FSVKTNIRSSTRDWKDHKFKWRKDPQDK | |
25 μL | |
Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
2214 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction