Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FcεR1α Armenian Hamster anti-Mouse, Alexa Fluor™ 488, Clone: MAR-1 (also known as MAR1), BD Pharmingen™

Catalog No. BDB567801 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567801 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB567801 Supplier BD Biosciences Supplier No. 567801
Add to Cart
Add to Cart
This item is not returnable. View return policy

Armenian Hamster Monoclonal Antibody

FcεR1α Monoclonal antibody specifically detects FcεR1α in Mouse samples. It is validated for Flow cytometry (Routinely Tested)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FcεR1α
Applications Flow Cytometry
Classification Monoclonal
Clone MAR-1 (also known as MAR1)
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias FceR1 alpha; Fcer1a; FcεRIα; High affinity IgE Receptor; MAR1
Host Species Armenian Hamster
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.