Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FcεR1α Armenian Hamster anti-Mouse, BUV496, Clone: MAR-1 (also known as MAR1), BD OptiBuild™

Catalog No. BDB751763 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751763 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751763 Supplier BD Biosciences Supplier No. 751763
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

FcεR1α Monoclonal antibody specifically detects FcεR1α in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FcεR1α
Applications Flow Cytometry
Classification Monoclonal
Clone MAR-1 (also known as MAR1)
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 496
Description Ms FceR1a BUV496 MAR-1 50 μg
Host Species Armenian Hamster
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity FceR1a
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG
Reactivity Mouse
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.