Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FcεR1α Mouse anti-Human, BUV737, Clone: AER-37 (also known as CRA-1, CRA1), BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 749338
Description
FcεR1α Monoclonal antibody specifically detects FcεR1α in Human samples. It is validated for Flow Cytometry
Specifications
FcεR1α | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
FCE1A; FCER1A; Fc-epsilon RI-alpha; FcERI; FceR1a; FcεRI | |
Recombinant Extracellular Domain of FcεR1α | |
RUO | |
Human | |
IgG2b κ |
Flow Cytometry | |
AER-37 (also known as CRA-1, CRA1) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction