Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fc Receptor Binding Inhibitor Polyclonal Antibody, eBioscience™
GREENER_CHOICE

Catalog No. 501122773
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2773 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-2773 Supplier Invitrogen™ Supplier No. 14916542
Add to Cart
Edge
Add to Cart

Polyclonal Antibody

Description: The Rhesus Fc Receptor Binding Inhibitor can be used to inhibit the non-specific Fc receptor (FcR)-mediated binding of mouse monoclonal antibodies used for flow cytometric analysis of rhesus tissue. FcR are expressed at varying levels in multiple cell lineages including high expression in myeloid cells, granulocytes, and B cells. The biological function of the FcR, including initiation of endocytosis, phagocytosis, and antigen presentation, is elicited upon binding of host-immunoglobulin. The extent to which mouse monoclonal antibodies will bind to rhesus FcR varies depending on the isotype of the monoclonal antibody. eBioscience testing indicates that mouse IgG2a isotype is blocked by Rhesus Fc Receptor Binding Inhibitor. Applications Reported: Rhesus Fc Receptor Binding Inhibitor has been reported for use in flow cytometric analysis. To inhibit the non-specific binding of mouse monoclonal antibodies add the Rhesus Fc Receptor Binding Inhibitor to samples and incubate on ice for 20 minutes. Without washing, proceed to stain with primary antibody. Applications Tested: Rhesus Fc Receptor Binding Inhibitor has been pre-titrated and tested by flow cytometric analysis of normal rhesus peripheral blood cells. This can be used at 5 μL (10 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/te...

Four different classes of Fcy receptors have been identified: FcyRI (CD64), FcyRII (CD32), and FcyRIII (CD16), and they are expressed at varying levels in multiple cell lineages including high expression in myeloid, granulocyte and B cells. The biological function of the FcyR, including initiation of endocytosis, phagocytosis and antigen presentation, is elicited upon binding of host-immunoglobulin. The extent to which mouse monoclonal antibodies will bind to human FcyR varies depending on the isotype of the monoclonal antibody. Furthermore, different monoclonal antibodies of the same isotype will display different binding to human FcyRs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc Receptor Binding Inhibitor
Applications Flow Cytometry
Classification Polyclonal
Concentration 5 μL/Test
Conjugate Unconjugated
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Alias Fc blocking; FcR BI; FcR Inhibitor
Host Species Not Available
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rhesus Macaque
Content And Storage 4°C
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.