Learn More
Fcμ Receptor Mouse anti-Mouse , Biotin, Clone: MM3, BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | Fcμ Receptor |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MM3 |
| Conjugate | Biotin |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Fcmr; FcmuR; Fcμ Receptor; FcμR; Faim3; Toso |
| Host Species | Mouse |
| Immunogen | Mouse FcμR Transfected Cell Line |
| Quantity | 0.5 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.