Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FcγRIIA (CD32) Mouse anti-Human, BV605, Clone: IV.3 (also known as IV3), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 755300
Description
FcγRIIA (CD32) Monoclonal antibody specifically detects FcγRIIA (CD32) in Human samples. It is validated for Flow cytometry
Specifications
FcγRIIA (CD32) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
2212 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2b κ |
Flow Cytometry | |
IV.3 (also known as IV3) | |
Brilliant Violet 605 | |
CD32a/FcγRIIa/FcgRIIA/FCGR2A; CD32b/FcγRIIb/FcgRIIB/FCGR2B | |
Human K562 leukemia cell line | |
50 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction