Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FcγRIIA (CD32) Mouse anti-Human, PE, Clone: IV.3 (also known as IV3), BD Pharmingen™
Mouse Monoclonal Antibody
$138.60 - $333.50
Specifications
Antigen | FcγRIIA (CD32) |
---|---|
Clone | IV.3 (also known as IV3) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
FcγRIIA (CD32) Monoclonal antibody specifically detects FcγRIIA (CD32) in Human samples. It is validated for Flow cytometry (Routinely Tested)Specifications
FcγRIIA (CD32) | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
CD32a/FcγRIIa/FcgRIIA/FCGR2A; CD32b/FcγRIIb/FcgRIIB/FCGR2B | |
Human K562 leukemia cell line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
IV.3 (also known as IV3) | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
2212 | |
IgG2b κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title