Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FcγRIV (CD16-2) Hamster anti-Mouse, Brilliant Violet 421, Clone: 9E9., BD Optibuild™

Catalog No. BDB742562 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742562 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742562 Supplier BD Biosciences Supplier No. 742562
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

The 9E9 monoclonal antibody specifically binds to Fc receptor, IgG, low affinity IV (FcγRIV), which is also known as CD16-2. FcγRIV is encoded by Fcgr4 /and belongs to the family of receptors for the Fc region of IgG (FcγRs), which also includes FcγRI (CD64), FcγRII (CD32), and FcγRIII (CD16) within the Ig superfamily.FcγRIVis expressed on monocytes, macrophages, dendritic cells, and neutrophils.FcγRIV, which serves as a ligand-biding subunit, requiresthe common Fcγ chain for expression and signaling. This complex serves as a cell activating receptor when bound by IgG2a or IgG2b. FcγRIV plays various roles in inflammation including neutrophil trafficking and mast cell degranulation.FcγRIVcan also function as a low-affinity IgE receptor and promote IgE-induced inflammation

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FcγRIV (CD16-2)
Classification Monoclonal
Clone 9E9.
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD16-2, Fcgr4, FcgRIV, Fc gammaRIV, FcgammaRIV, Fcgr3a, Fcrl3
Host Species Armenian Hamster
Immunogen Mouse FcγRIV Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.