Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCAMR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112125 Supplier Invitrogen™ Supplier No. PA5112125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FCAMR Polyclonal Antibody for Western Blot, IHC (P)

Functions as a receptor for the Fc fragment of IgA and IgM. Binds IgA and IgM with high affinity and mediates their endocytosis. May function in the immune response to microbes mediated by IgA and IgM. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCAMR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene FCAMR
Gene Accession No. Q2TB54
Gene Alias CD351; fc alpha/mu receptor; Fc fragment of IgA and IgM receptor; Fc IgA/IgM receptor; Fc receptor, IgA, IgM, high affinity; Fca/m receptor; FCA/MR; Fcamr; FKSG87; high affinity immunoglobulin alpha and immunoglobulin mu Fc receptor; immunity related factor; mFcamR; receptor for Fc fragment of IgA and IgM
Gene Symbols FCAMR
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the N-terminus of the Mouse FCAMR/CD351.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64435
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.