Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCAR/CD89 Antibody (013), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290841AF700
Description
FCAR/CD89 Monoclonal specifically detects FCAR/CD89 in Rat samples. It is validated for ELISA.Specifications
FCAR/CD89 | |
Monoclonal | |
Alexa Fluor 700 | |
CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat FCAR/CD89 (NP_973721.1; Met1-Asn228). | |
0.1 mL | |
Immunology | |
2204 | |
Rat | |
Purified |
ELISA | |
13 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Rat FCGR1a | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction