Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCAR/CD89 Antibody (14A8 (14.1)), Alexa Fluor™ 750 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320161AF750
Description
FCAR/CD89 Monoclonal antibody specifically detects FCAR/CD89 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
FCAR/CD89 | |
Monoclonal | |
Alexa Fluor 750 | |
CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor | |
The original antibody was generated by immunizing HCO12 trangene containing mice with recombinant soluble CD89 antigen. | |
0.1 mL | |
Immunology | |
2204 | |
Store at 4°C in the dark. | |
IgG κ |
Flow Cytometry, ELISA | |
14A8 (14.1) | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction