Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCAR/CD89 Antibody (MIP8a) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $501.50
Specifications
Antigen | FCAR/CD89 |
---|---|
Clone | MIP8a |
Concentration | 1.0 mg/mL |
Dilution | Western Blot, Flow Cytometry 1:50-1:100, ELISA, Immunoprecipitation |
Applications | ELISA, Western Blot, Immunoprecipitation, Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017545
![]() |
Novus Biologicals
NB100632660.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10063266
![]() |
Novus Biologicals
NB10063266 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
FCAR/CD89 Monoclonal specifically detects FCAR/CD89 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation.Specifications
FCAR/CD89 | |
1.0 mg/mL | |
ELISA, Western Blot, Immunoprecipitation, Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor | |
FCAR | |
IgG1 | |
Protein A or G purified | |
NB100-63266 recognizes the human CD89 cell surface antigen, a 50-75kD cell surface glycoprotein that is also known as the IgA receptor (Fc alpha R). CD89 is expressed by peripheral blood monocytes and neutrophils. Clone MIP8a blocks binding of IgA to the Fc alpha R, and also inhibits neutrophil phagocytosis of IgA complexes. |
MIP8a | |
Western Blot, Flow Cytometry 1:50-1:100, ELISA, Immunoprecipitation | |
Monoclonal | |
Purified | |
RUO | |
P24071 | |
2204 | |
Recombinant soluble human Fc alpha R | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title