Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCAR/CD89 Antibody (MIP8a), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063266APCCY7
Description
FCAR/CD89 Monoclonal specifically detects FCAR/CD89 in Human samples. It is validated for Flow Cytometry.Specifications
FCAR/CD89 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
FCAR | |
Recombinant soluble human Fc alpha R | |
0.1 mL | |
Primary | |
NB100-63266 recognizes the human CD89 cell surface antigen, a 50-75kD cell surface glycoprotein that is also known as the IgA receptor (Fc alpha R). CD89 is expressed by peripheral blood monocytes and neutrophils. Clone MIP8a blocks binding of IgA to the Fc alpha R, and also inhibits neutrophil phagocytosis of IgA complexes. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
MIP8a | |
Flow Cytometry | |
CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor | |
Mouse | |
Protein A or G purified | |
Immunology | |
2204.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction