Learn More
Invitrogen™ FCAR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121219
Description
Positive test controls include: 293T. The target is usually found in the following locations: Cell membrane, Secreted, Single-pass type I membrane protein. Immunogen sequence: LMIIKNSTYR EIGRRLKFWN ETDPEFVIDH MDANKAGRYQ CQYRIGHYRF RYSDTLELVV TGLYGKPFLS ADRGLVLMPG ENISLTCSSA HIPFDRFSLA KEGELSLPQH QSGEHPANFS LGPVDLNVSG IYRCYGWYNR SPYLWSFPSN ALELVVTDSI HQDYTTQNLI RMAVAGLVLV ALLAILVENW HSHTALNKEA SADVAEPSWS QQMCQPGLTF ARTPSVCK.
This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
| FCAR | |
| Polyclonal | |
| Unconjugated | |
| Fcar | |
| CD89; CTB-61M7.2; Fc alpha receptor; Fc fragment of IgA receptor; Fc fragment of IgA, receptor for; FcalphaRI; FCAR; IgA Fc receptor; immunoglobulin A Fc receptor; immunoglobulin A Fc receptor transcript variant 1; immunoglobulin A Fc receptor transcript variant 3; immunoglobulin A Fc receptor transcript variant 7; immunoglobulin A Fc receptor transcript variant 9; immunoglobulin A Fc receptor transcript variant deltaEC1; immunoglobulin A Fc receptor transcript variant deltaEC1 delta TM/C; immunoglobulin alpha Fc receptor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2204 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.66 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P24071 | |
| Fcar | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-287 of human FCAR (NP_001991.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.