Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FCAR Recombinant Rabbit Monoclonal Antibody (6)
GREENER_CHOICE

Catalog No. PIMA530407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530407 Supplier Invitrogen™ Supplier No. MA530407
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CD89/FCAR.

This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCAR
Applications ELISA
Classification Recombinant Monoclonal
Clone 6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Fcar
Gene Accession No. P24071
Gene Alias CD89; CTB-61M7.2; Fc alpha receptor; Fc fragment of IgA receptor; Fc fragment of IgA, receptor for; FcalphaRI; FCAR; IgA Fc receptor; immunoglobulin A Fc receptor; immunoglobulin A Fc receptor transcript variant 1; immunoglobulin A Fc receptor transcript variant 3; immunoglobulin A Fc receptor transcript variant 7; immunoglobulin A Fc receptor transcript variant 9; immunoglobulin A Fc receptor transcript variant deltaEC1; immunoglobulin A Fc receptor transcript variant deltaEC1 delta TM/C; immunoglobulin alpha Fc receptor
Gene Symbols Fcar
Host Species Rabbit
Immunogen Recombinant Human CD89 protein (Met1-Asn227).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2204
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.