Learn More
Invitrogen™ FceR1 alpha Monoclonal Antibody (1F2A9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA517074
Description
MA5-17074 targets FCER1A in indirect ELISA, FACS and WB applications and shows reactivity with Human samples. The MA5-17074 immunogen is purified recombinant fragment of human FCER1A (amino acids:42-103) expressed in E. Coli. MA5-17074 detects FCER1A which has a predicted molecular weight of approximately 29.6kDa.
Fc epsilon RI alpha, also known as FceR1 alpha, is a subunit of the high-affinity receptor for IgE, primarily expressed on mast cells and basophils. Its expression is upregulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits, which are essential for signal transduction. The beta and gamma subunits contain immunoreceptor tyrosine-based activation motifs (ITAMs). The Fc epsilon RI complex plays a crucial role in triggering IgE-mediated allergic reactions. When allergen-bound IgE molecules bring together two or more high-affinity IgE receptors, it leads to the release of mediators such as histamine, which are responsible for allergy symptoms. This process couples allergens and mast cells to initiate inflammatory responses characteristic of allergic disorders like hay fever and asthma. The release of histamine and proteases also leads to the synthesis of prostaglandins and leukotrienes, which are potent effectors of the hypersensitivity response.
Specifications
FceR1 alpha | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P12319, P20489 | |
FCER1A | |
Purified recombinant fragment of human FCER1A (amino acids:42-103) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
1F2A9 | |
Unconjugated | |
FCER1A | |
alpha polypeptide; fc epsilon r1; Fc epsilon receptor Ia; Fc epsilon RI alpha-chain; Fc fragment of IgE receptor Ia; Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide; Fc IgE receptor, alpha polypeptide; Fc receptor, IgE, high affinity I, alpha polypeptide; Fce1a; Fc-epsilon RI-alpha; fcepsilonri; FCER1A; FcERI; Fcr-5; high affinity immunoglobulin epsilon receptor alpha-subunit; high affinity immunoglobulin epsilon receptor subunit alpha; IgE Fc receptor subunit alpha; Iger01; immunoglobulin E receptor, high-affinity, of mast cells, alpha polypeptide; RATIGER01; RP11-550P17.3 | |
Mouse | |
Protein G | |
RUO | |
14125, 2205 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.