Learn More
Invitrogen™ FceR1 alpha Monoclonal Antibody (CRA2 (AER24)), Biotin

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA14999
Description
MA1-4999 detects Fc epsilonR1 alpha (Biotin) from human samples. The CRA2 (AER24) monoclonal antibody reacts with the Fc epsilonR1 alpha subunit on a region that overlaps the region of the IgE binding site, thus it competes with IgE for the receptor bindi MA1-4999 has been successfully used in Western blot, flow cytometry, and immunohistochemistry applications. The MA1-4999 immunogen is region of Fc epsilonR1 alpha which overlaps with the IgE binding site; Swiss protein P12319. The epitope has been mapped to amino acids 110-197.
Fc epsilon RI alpha, also known as FceR1 alpha, is a subunit of the high-affinity receptor for IgE, primarily expressed on mast cells and basophils. Its expression is upregulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits, which are essential for signal transduction. The beta and gamma subunits contain immunoreceptor tyrosine-based activation motifs (ITAMs). The Fc epsilon RI complex plays a crucial role in triggering IgE-mediated allergic reactions. When allergen-bound IgE molecules bring together two or more high-affinity IgE receptors, it leads to the release of mediators such as histamine, which are responsible for allergy symptoms. This process couples allergens and mast cells to initiate inflammatory responses characteristic of allergic disorders like hay fever and asthma. The release of histamine and proteases also leads to the synthesis of prostaglandins and leukotrienes, which are potent effectors of the hypersensitivity response.
Specifications
FceR1 alpha | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and no preservative; pH 7.4 | |
P12319 | |
FCER1A | |
Region of FceR1 alpha (aa 26-197). | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
CRA2 (AER24) | |
Biotin | |
FCER1A | |
alpha polypeptide; fc epsilon r1; Fc epsilon receptor Ia; Fc epsilon RI alpha-chain; Fc fragment of IgE receptor Ia; Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide; Fc IgE receptor, alpha polypeptide; Fc receptor, IgE, high affinity I, alpha polypeptide; Fce1a; Fc-epsilon RI-alpha; fcepsilonri; FCER1A; FcERI; Fcr-5; high affinity immunoglobulin epsilon receptor alpha-subunit; high affinity immunoglobulin epsilon receptor subunit alpha; IgE Fc receptor subunit alpha; Iger01; immunoglobulin E receptor, high-affinity, of mast cells, alpha polypeptide; RATIGER01; RP11-550P17.3 | |
Mouse | |
IgG fraction | |
RUO | |
2205 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.