Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FceR1 alpha Monoclonal Antibody (MAR-1), FITC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 11589882
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-589-882 100 μg
50-991-1 50 μg
11-589-885 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-589-882 Supplier Invitrogen™ Supplier No. 11589882
Add to Cart
Edge
Add to Cart

Armenian Hamster Monoclonal Antibody

Description: The MAR-1 monoclonal antibody reacts with the Fc epsilon Receptor I alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI alpha is expressed on mast and basophil cells and is up-regulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions. Applications Reported: The MAR-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: The MAR-1 antibody has been tested by flow cytometric analysis of the MC/9 cell line (a mouse mast cell line). This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Fc epsilon RI alpha, also known as FceR1 alpha, is a subunit of the high-affinity receptor for IgE, primarily expressed on mast cells and basophils. Its expression is upregulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits, which are essential for signal transduction. The beta and gamma subunits contain immunoreceptor tyrosine-based activation motifs (ITAMs). The Fc epsilon RI complex plays a crucial role in triggering IgE-mediated allergic reactions. When allergen-bound IgE molecules bring together two or more high-affinity IgE receptors, it leads to the release of mediators such as histamine, which are responsible for allergy symptoms. This process couples allergens and mast cells to initiate inflammatory responses characteristic of allergic disorders like hay fever and asthma. The release of histamine and proteases also leads to the synthesis of prostaglandins and leukotrienes, which are potent effectors of the hypersensitivity response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FceR1 alpha
Applications Flow Cytometry
Classification Monoclonal
Clone MAR-1
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene FCER1A
Gene Accession No. P20489
Gene Alias alpha polypeptide; fc epsilon r1; Fc epsilon receptor Ia; Fc epsilon RI alpha-chain; Fc fragment of IgE receptor Ia; Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide; Fc IgE receptor, alpha polypeptide; Fc receptor, IgE, high affinity I, alpha polypeptide; Fce1a; Fc-epsilon RI-alpha; fcepsilonri; FCER1A; FcERI; Fcr-5; high affinity immunoglobulin epsilon receptor alpha-subunit; high affinity immunoglobulin epsilon receptor subunit alpha; IgE Fc receptor subunit alpha; Iger01; immunoglobulin E receptor, high-affinity, of mast cells, alpha polypeptide; RATIGER01; RP11-550P17.3
Gene Symbols FCER1A
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14125
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.